Disoprofol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2078-54-8
:
Formula:C₁₂H₁₈O
MDL Number:MFCD00008885
:178.27
Merck Index:13,07927
Boiling Pt:256 °C (1013 hPa)
Melting Pt:18 °C
Density:1.032 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:218-206-6