Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Disoprofol
Disoprofol
Disoprofol
MOLE66803840-100MG
:
Molekula
:
Disoprofol
MOLE66803840-100MG
:
Molekula
:
Pk:
100 mg
Seq:
2440
:
:
178.27
:
13,07927
:
256 °C (1013 hPa)
:
18 °C
:
1.032 g/cm³ (20 °C)
:
218-206-6
:
2078-54-8
:
C₁₂H₁₈O
:
MFCD00008885