Oxybenzone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:131-57-7
:
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00008387
:228.25 g/mol
Merck Index:12,07088
Boiling Pt:370 °C (1013 hPa)
Melting Pt:65 °C
Density:1.3397 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
EINECS:205-031-5