Oxybenzone 98%
:131-57-7
:
:C₁₄H₁₂O₃
:MFCD00008387
:228.25 g/mol
:12,07088
:150…160 °C (5 mmHg)
:>62.5 °C
:1,3397 g/cm³ (25 °C)
:>100 °C
:Ambient
:205-031-5
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Melting point:≥62.0 °C
- GC:≥97.5 %
- Specific extinction E (1%/1cm):≥400 (321 to 327 nm, methanol)