4-Heptylphenol
Catalog # EHERC14136500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:192.3 g/mol
Boiling Pt:298 °C (1013 hPa)
Melting Pt:26…28 °C
Density:0.939 g/cm³ (20 °C)
EINECS:217-862-0
UN:2430
ADR:8,III
CAS Number:1987-50-4
Formula:C₁₃H₂₀O
MDL Number:MFCD00041751