Order Entry
Germany
ContactUsLinkComponent
 
4'-Hydroxy-3'-methoxyacetophenone, Millipore®
Catalog # 178385-1
CAS Number:  
4'-Hydroxy-3'-methoxyacetophenone, Millipore®
Catalog # 178385-1
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.18 g/mol
Merck Index:13,00746
Boiling Pt:295 °C (1013 hPa)
Melting Pt:115 °C
EINECS:207-854-5
CAS Number:498-02-2
Formula:C₉H₁₀O₃
MDL Number:MFCD00008747