4'-Methoxyacetophenone
Catalog # EHERC15059000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:150.18 g/mol
Boiling Pt:152 °C (1013 hPa)
Melting Pt:36…38 °C
Density:1,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-815-9
:GHS07
CAS Number:100-06-1
Formula:C₉H₁₀O₂
MDL Number:MFCD00008745