Order Entry
Germany
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

100 mL
Catalog # PANBP10-036100
Marketsource
Supplier:   PAN BIOTECH
Trypsin
Catalog # PANBP10-036100
Supplier:  PAN BIOTECH
Marketsource
CAS Number:  
Trypsin

100 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents

Trypsin is a porcine pancreas-derived enzyme that is commonly usedin dissociation of anchorage dependent mammalian cells and tissues.

  • Stability: two years from production

The concentration of trypsin is necessary to dislodge cells from their substrate is dependent on the cell type and age of the culture. EDTA, a chelating reagent is added to trypsin solution to enhance enzymatic activity by neutralising calcium and magnesium ions that enhance cell to cell adhesion and obscure peptide bonds on which trypsin acts.

Specifications

Specifications

  • Pk:
    100 mL
  • Cat. No.:
    PANBP10-036100
  • Form:
    Solution
  • Storage conditions:
    −20 °C
  • Concentration:
    0,25% trypsin/0,53 mM EDTA in HBBS, without calcium and magnesium, with phenol red
  • Enzyme name:
    Trypsin

Specifications