5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Catalog # CAYM703012-1
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:396.36 g/mol
Melting Pt:240…244 °C
EINECS:200-714-4
:GHS07
Storage Temperature:Ambient
CAS Number:69-78-3
Formula:C₁₄H₈N₂O₈S₂
MDL Number:MFCD00007140