About this item
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life.
- RIPEL™ technology prevents liquids and oil from making skin contact keeping hands safe and dry
- Thumb crotch reinforcement technology for up to 12x the durability of the inherently weak thumb crotch region*
- ANSELL GRIP™ technology delivers superior grip performance for safer handling of wet or oily parts
- FORTIX™ technology provides extended glove life and high abrasion resistance
- ZONZ™ comfort fit technology provides dexterity and comfort
The 18 gauge liner featuring Dyneema® Diamond Technology provides high dexterity and comfort. The foam nitrile coating and integrated thumb crotch reinforcement (TCR) offer long-lasting protection and 12x better wear life than unreinforced gloves based on field tests. Foam nitrile ¾ coating provides liquid repellence. A silicone free design insures full product protection.
Recommended for: Inspection, selecting, checking parts; assembly & inspection of components; fastening components; chassis assembly, body trimming; adjusting parts; delivery of parts to production line; equipment repair & maintenance; engine transmission and repair; oil, fluids and filter change; inspection (picking, checking and marking); feeding the press and removing spare parts from it; adjusting panels parts components; press shop operations.
*compared to unreinforced TCR competitor gloves.
Oeko-Tex; REACH compliant.
Specifications
- Catalog No:
- ANSE11-937/11
- ANSE11-937/8
- ANSE11-937/6
- ANSE11-937/7
- ANSE11-937/9
- ANSE11-937/10
- Model:
- 11-937
- 11-937
- 11-937
- 11-937
- 11-937
- 11-937
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Dyneema®
- Dyneema®
- Dyneema®
- Dyneema®
- Dyneema®
- Dyneema®
- Finish:
- Foam nitrile ¾ coated
- Foam nitrile ¾ coated
- Foam nitrile ¾ coated
- Foam nitrile ¾ coated
- Foam nitrile ¾ coated
- Foam nitrile ¾ coated
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- Anti-static:
- +
- +
- +
- +
- +
- +