Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4-Trimethoxyphenylboronic acid
2,3,4-Trimethoxyphenylboronic acid 95+%
2,3,4-Trimethoxyphenylboronic acid 95+%
2,3,4-Trimethoxyphenylboronic acid 95+%
Catalog # CHMP902615.POR
Supplier:
CHEMPUR
CAS Number:
Expand image
2,3,4-Trimethoxyphenylboronic acid 95+%
Catalog # CHMP902615.POR
Supplier:
CHEMPUR
CAS Number:
Specifications
Pk:
1
Seq:
0001
Specifications
About this item
Product Details
:
:
212.01 g/mol
CAS Number:
118062-05-8
Formula:
C₉H₁₃BO₅
MDL Number:
MFCD01318183
Product Details