Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®
Certificates
undefined
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    White to Off-White
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Colorless to Faint Yellow
  • Solubility (Turbidity)
    50 mg/mL, H2O:
    Clear
  • Viscosity
    4 % Aqueous solution at 20 deg C (Hoeppler Method):
    11.6 - 15.4 cps
  • Hydrolysis:
    99.2 - 99.7 mole%
  • Recommended Retest Period:
    4 years

Specification Test Results