4-Phenylmorpholine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:92-53-5
:
Formula:C₁₀H₁₃NO
MDL Number:MFCD00006166
Boiling Pt:268 °C (1013 hPa)
Melting Pt:51…54 °C
Density:1.058 g/cm³ (20 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:163.22 g/mol
Storage Temperature:Ambient
EINECS:202-164-0
UN:2811
:GHS06
Specification Test Results
- Melting Point:48 - 51 °C
- Molar Mass:163.22 g/mol
- Boiling Point:260 °C (1013 hPa)
- Flash Point:124 °C
- Density:1.053 g/cm3 (60 °C)
- Ignition Point:410 °C