Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MOPS-Na
MOPS-Na, Sigma-Aldrich®
MOPS-Na, Sigma-Aldrich®
MOPS-Na, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
MOPS-Na, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
71119-22-7
:
Formula:
C₇H₁₄NNaO₄S
MDL Number:
MFCD00064350
:
231.25 g/mol
:
M-Clarity™ quality level = MQ400
Storage Temperature:
Ambient
EINECS:
428-420-3