Order Entry
Germany
Orders LinkContactUsLinkComponent
MOPS-Na

MOPS-Na

MOPS-Na
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
  • CAS Number:71119-22-7
  • Formula:C₇H₁₄NNaO₄S
  • MDL Number:MFCD00064350
  • Molecular Weight:231.25 g/mol
  • Storage Temperature:Ambient
  • EINECS:428-420-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.