β-(N-Morpholino)ethanesulphonic acid (MES)
Supplier: Molekula
Certificates
About this item
CAS Number:4432-31-9
:
Formula:C₆H₁₃NO₄S
MDL Number:MFCD00006181
:195.24 g/mol
Merck Index:13,05929
Melting Pt:316 °C
Storage Temperature:Ambient
EINECS:224-632-3