
Malonaldehyde dianilide hydrochloride ≥98.0% (by HPLC, titration analysis)
TCIAM1288-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
:
:258.75 g/mol
:248-862-9
:GHS07
:28140-60-5
:50328-50-2
:C₁₅H₁₄N₂
:MFCD00054634