
Imiquimod ≥98,0% (durch HPLC, titrimetrische Analyse)
Supplier: TCI
Certificates
About this item
CAS Number:99011-02-6
:
Formula:C₁₄H₁₆N₄
MDL Number:MFCD00866946
:240.31 g/mol
Merck Index:14,04922
Melting Pt:294°C
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %