
(4-Hydroxyphenyl)phosphonic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:33795-18-5
:
Formula:C₆H₇O₄P
MDL Number:MFCD01112120
:174.09 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %