Order Entry
Germany
ContactUsLinkComponent
Itaconic acid
Itaconic acid
Catalog # MOLE10565984-1KG
Supplier:  Molekula
CAS Number:  
Itaconic acid
Catalog # MOLE10565984-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :130.1 g/mol
Merck Index:13,05264
Boiling Pt:381 °C (1013 hPa)
Melting Pt:162…167 °C
Density:1.573 g/cm³ (20 °C)
EINECS:202-599-6
 :GHS05
CAS Number:97-65-4
Formula:C₅H₆O₄
MDL Number:MFCD00004260