Order Entry
Germany
ContactUsLinkComponent
Itaconic acid for synthesis, Sigma-Aldrich®
Corrosive
Itaconic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Itaconic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    83 g/l (20 °C)
  • Melting Point:
    162 - 167 °C
  • Molar Mass:
    130.1 g/mol
  • Boiling Point:
    268 °C (decomposition)
  • Density:
    1.632 g/cm3 (20 °C)
  • pH:
    2 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    430 °C

Specification Test Results

Related Information