Itaconic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-65-4
:
Formula:C₅H₆O₄
MDL Number:MFCD00004260
:130.1 g/mol
Merck Index:14,5242
Boiling Pt:268 °C
Melting Pt:165…169 °C
Density:1.630
EINECS:202-599-6
:GHS07
Storage Temperature:Ambient