1-Pentanesulphonic acid sodium salt ≥98% for ion pair chromatography, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:22767-49-3
:
Formula:C₅H₁₁O₃SNa
MDL Number:MFCD00007541
:174.2 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:245-208-4
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 98.0 %
- Assay (as C₅H₁₁SO₃Na) (by acidimetry):≥ 98.0 %
- UV Absorbance of 0.25 M Solution - 250 nm:≤ 0.050
- UV Absorbance of 0.25 M Solution - 240 nm:≤ 0.050
- UV Absorbance of 0.25 M Solution - 230 nm:≤ 0.050
- UV Absorbance of 0.25 M Solution - 220 nm:≤ 0.060
- UV Absorbance of 0.25 M Solution - 210 nm:≤ 0.080
- UV Absorbance of 0.25 M Solution - 200 nm:≤ 0.200