Glycerine
KRAE900235
:
- Pk:250 kg
- Seq:4748
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
:
:92.09 g/mol
:13,04497
:290 °C (1013 hPa)
:18.6 °C
:1.26 g/cm³ (20 °C)
:177 °C
:Ambient
:200-289-5
:56-81-5
:HOCH₂CH(OH)CH₂OH
:MFCD00004722