Acetal, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-57-7
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00009243
:118.18 g/mol
Merck Index:13,00040
Boiling Pt:102.7 °C (1013 hPa)
Melting Pt:–100 °C
Density:0.83 g/cm³ (20 °C)
EINECS:203-310-6
UN:1088
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:–21 °C
:GHS02
:GHS07
Storage Temperature:Refrigerator