Acetal ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105-57-7
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00009243
:118.18 g/mol
Merck Index:14,38
Boiling Pt:101…102 °C
Melting Pt:–100 °C
Density:0.831
Flash Pt:–21 °C (–6 °F)
Storage Temperature:Refrigerator
EINECS:203-310-6
UN:1088
ADR:3,II
:GHS02
:GHS07