Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethylenetriaminepentaacetic acid
Diethylenetriaminepentaacetic acid, Sigma-Aldrich®
Diethylenetriaminepentaacetic acid, Sigma-Aldrich®
:
Merck
Diethylenetriaminepentaacetic acid, Sigma-Aldrich®
:
Merck
:
67-43-6
:
:
C₁₄H₂₃N₃O₁₀
:
MFCD00004289
:
13,07202
:
220…222 °C
:
1.558 g/cm³ (20 °C)
:
Ambient
:
200-652-8
:
3077
:
9,III
:
393.35 g/mol
:
GHS07