Diethylenetriaminepentaacetic acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-43-6
:
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289
Merck Index:13,07202
Melting Pt:220…222 °C
Density:1.558 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:9,III
:M-Clarity™ quality level = MQ100
:393.35 g/mol
EINECS:200-652-8
UN:3077
:GHS08
:GHS07
Specification Test Results
- Solubility:4.8 g/l (25 °C)
- Melting Point:220 - 222 °C decomposes
- Molar Mass:393.34 g/mol
- Bulk Density:580 kg/m3
- Density:1.56 g/cm3
- pH:2 - 3 (H2O, 20 °C) (saturated solution)