Diethylenetriaminepentaacetic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-43-6
:
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289
Merck Index:14,7125
Melting Pt:∼ 220 °C (dec.)
Density:1.56
EINECS:200-652-8
UN:3077
ADR:9,III
:393.35 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient