Diethylenetriaminepentaacetic acid 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-43-6
:
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289
Merck Index:13,07202
Melting Pt:220 °C (dec.)
Density:1.56 g/cm³
Flash Pt:200 °C
EINECS:200-652-8
ADR:9,III
:393.35 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration excess NaOH:≥98.0 %