Calcium acetate
25 kg
Catalog # KRAE900152-9025
Supplier: KRAEMER MARTIN
Jump to:
Product Details & Documents
Synonyms: Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt
- CAS Number:62-54-4
- Formula:(H₃CCOO)₂Ca
- MDL Number:MFCD00150019
- Molecular Weight:158.17 g/mol
- Merck Index:12,01683
- Density:1.509 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-540-9
Specifications
Specifications
- Pk:25 kg



