
2-Methoxy-1-methylethyl acetate (PGMEA)
Catalog # KRAE17-194.5000
Supplier: KRAEMER MARTIN
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:132.16 g/mol
Boiling Pt:145.8 °C (1013 hPa)
Melting Pt:–67 °C
Density:0.968 g/cm³ (20 °C)
Flash Pt:45 °C
EINECS:203-603-9
UN:3272
ADR:3,III
:GHS02
CAS Number:108-65-6
Formula:C₆H₁₂O₃
MDL Number:MFCD00038500