Triacetin (glycerol triacetate)
Catalog # KRAE900670.9230
Supplier: KRAEMER MARTIN
CAS Number:
Specifications
- Pk:230 kg
- Seq:4732
Specifications
About this item
:
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716