Calcium pantothenate USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:137-08-6
:
Formula:C₁₈H₃₂CaN₂O₁₀
MDL Number:MFCD00002766
:476.54 g/mol
Melting Pt:74 °C
Storage Temperature:Ambient
EINECS:205-278-9
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- Identification A:Passes Test
- Identification B:Passes Test
- Identification C (Specific Rotation [Å]²⁵ ^D (+)):25.0 - 27.5 Degree
- Alkalinity:Passes Test
- Loss on Drying:≤5.0 %
- Assay (C₁₈H₃₂CaN₂O₁₀)(dried basis):98.0 - 102.0 %
- Content of Calcium (Ca)(dried basis):8.2 - 8.6 %
- Solution Test:Passes Test