
Adenine sulphate dihydrate ≥98.0% (by HPLC, titration analysis)
: TCI
:6509-19-9
:MFCD00150149
:(C₅H₅N₅)₂·H₂SO₄·2H₂O
:MFCD00213655
:404.37 g/mol
:14,00152
:206-286-5
:GHS07
- Appearance:White to Light yellow powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on dried substance)
- Solubility in hot Water:almost transparency
- Drying loss:8.0 to 10.0 %