Order Entry
Germany
ContactUsLinkComponent
Cobalt(II) perchlorate hexahydrate, Sigma-Aldrich®
Cobalt(II) perchlorate hexahydrate, Sigma-Aldrich®
Catalog # SIAL401404-25G
Supplier:  Merck
CAS Number:  
Cobalt(II) perchlorate hexahydrate, Sigma-Aldrich®
Catalog # SIAL401404-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :365.93 g/mol
ADR:5.1,II
 :M-Clarity™ quality level = MQ100
EINECS:236-653-5
UN:1481
CAS Number:13478-33-6
Formula:Cl₂CoO₈·6H₂O
MDL Number:MFCD00150207