Zinc perchlorate hexahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-64-6
:
Formula:Cl₂O₈Zn·6H₂O
MDL Number:MFCD00150370
:372.38 g/mol
Merck Index:14,10148
Melting Pt:106 °C
Density:2.25
Storage Temperature:Ambient
EINECS:237-122-0
UN:1481
ADR:5.1,II
:GHS03
:GHS07