(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Catalog # FLUO001472-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:132.16 g/mol
Merck Index:12,05232
Boiling Pt:72 °C (1013 hPa)
Density:1.07 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:405-320-8
CAS Number:14347-78-5
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213