(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14347-78-5
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213
:132.16 g/mol
Merck Index:12,05232
Boiling Pt:72…73 °C (8 mmHg)
Density:1.06 g/cm³
Flash Pt:80 °C
Storage Temperature:Refrigerator
EINECS:405-320-8
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4360 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:-13° to -15° (20°C, 589 nm) (neat)