Order Entry
Germany
ContactUsLinkComponent
 
Calcium hydrogen orthophosphate dihydrate
Catalog # CSLO2142.0250
Supplier:  CAESAR LORENTZ
CAS Number:  
Calcium hydrogen orthophosphate dihydrate
Catalog # CSLO2142.0250
Supplier:  CAESAR LORENTZ
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :172.09 g/mol
Merck Index:13,01699
Melting Pt:36 °C
Density:2.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-826-1
CAS Number:7789-77-7
Formula:HCaO₄P·2H₂O
MDL Number:MFCD00149621