L(+)-Lysine monohydrochloride
Catalog # MOLE11195192-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Melting Pt:263…264 °C
:182.65 g/mol
Storage Temperature:Ambient
EINECS:211-519-9
CAS Number:657-27-2
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564