Order Entry
Germany
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥99%, Ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥99%, Ultrapure
Catalog # E169-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥99%, Ultrapure
Catalog # E169-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Abs.@260 nm (1 M, Water):
    <0.02
  • Abs.@280 nm (1 M, Water):
    <0.02
  • DNase:
    none detected
  • Appearance:
    White crystalline powder
  • Moisture (KF):
    <10%
  • pKa @25 °C:
    5.9 – 6.3
  • Protease:
    none detected
  • Purity (Dry Basis,Titration):
    >99%
  • RNase:
    none detected
  • Solubility (1 M, Water):
    Pass

Specification Test Results

About this item

 :
 :213.25 g/mol
Merck Index:13,05929
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:224-632-3
CAS Number:145224-94-8
Formula:C₆H₁₃NO₄S·1H₂O
MDL Number:MFCD00149409