Order Entry
Germany
ContactUsLinkComponent
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Catalog # BACE4030835.0500
Supplier:  BACHEM BIOCHEMICA
CAS Number:  
L(+)-Lysine monohydrochloride
Catalog # BACE4030835.0500
Supplier:  BACHEM BIOCHEMICA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Melting Pt:263…264 °C
 :182.65 g/mol
Storage Temperature:Ambient
EINECS:211-519-9
CAS Number:657-27-2
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564