Order Entry
Germany
ContactUsLinkComponent
Environment
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Environment
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Titer (N):
    0.0997 - 0.1003
  • Chloride and Chlorate (as Cl):
    max. 10 ppm
  • Sulfate (SO4):
    max. 20 ppm

Specification Test Results

Related Information