Foetal bovine serum, delipidised, EU approved
Catalog # PANBP30-3402
Supplier: PAN BIOTECH
Specifications
- Sterility:Sterile
- Description:Foetal bovine serum, EU approved, delipidised
- Seq:02
- Pk:500 mL
Specifications
About this item
In this delipidised serum, lipids are removed from serum by affinity chromatography.
- Sterile solution
- Store at ‒20 °C
Applications include: Lipid metabolism studies, arteriosclerosis research.