Order Entry
Germany
ContactUsLinkComponent
 
Nickel standard solution, 10000 mg/L Ni in dil. nitric acid (from Ni(NO₃)₂) Certipur® standard for ICP, Supelco®
  1.70382.0100
 :  Merck
 :  
Nickel standard solution, 10000 mg/L Ni in dil. nitric acid (from Ni(NO₃)₂) Certipur® standard for ICP, Supelco®
  1.70382.0100
 :  Merck
 :  

 

  • Concentration:
    10000mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Nickel
  • Symbol:
    Ni

 

 

 : Traceable to SRM from NIST.