
Tetrapropylammonium hydroxide 40% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4499-86-9
:
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360
:203.37 g/mol
Boiling Pt:102 °C
Density:0.99
Storage Temperature:Ambient
EINECS:224-800-6
UN:3267
ADR:8,II
:GHS05