
Tetrapropylammonium hydroxide 10% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4499-86-9
:
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360
:203.37 g/mol
Storage Temperature:Refrigerator
EINECS:224-800-6
UN:3267
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HClO4:9 to 11 %
- Refractive index:1.3950 to 1.4000 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:≤1.1 % Triethylamine