Order Entry
Germany
ContactUsLinkComponent
Sodium permanganate 40% in aqueous solution, Sigma-Aldrich®
Sodium permanganate 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL519073-500ML
Supplier:  Merck
CAS Number:  
Sodium permanganate 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL519073-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :141.93 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:233-251-1
CAS Number:10101-50-5
Formula:MnNaO₄