Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hypochlorite
Sodium hypochlorite (13% Cl₂) in aqueous solution
Sodium hypochlorite (13% Cl₂) in aqueous solution
:
Bernd Kraft
Sodium hypochlorite (13% Cl₂) in aqueous solution
:
Bernd Kraft
:
7681-52-9
:
:
ClNaO
:
MFCD00011120
:
74.44 g/mol
:
111 °C (1013 hPa)
:
–30…–20 °C
:
1.22…1.25 g/cm³ (20 °C)
:
Refrigerator
:
231-668-3
:
1791
:
8,II
:
GHS05
:
GHS09